Invitrogen
Manufacturer Code:PA116948
Catalog # 01-950-177
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide between residues 180-232 of human VEGFA protein sequence. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: vascular endothelial growth factor; vascular endothelial growth factor (VEGF) precursor; vascular endothelial growth factor 164; vascular endothelial growth factor 165b; vascular endothelial growth factor 188; Vascular endothelial growth factor A; Vascular endothelial growth factor A antibody Vascular endothelial growth factor A precursor antibody Vascular permeability factor antibody VPF ant; vascular endothelial growth factor A121; vascular endothelial growth factor A165; Vascular permeability factor; VEGF; VEGF-A; VPF
Gene Aliases: eVEGF120; eVEGF164; MVCD1; VEGF; VEGF-A; VEGF164; VEGFA; VPF
UniProt ID: (Bovine) P15691, (Human) P15692, (Chicken) P67964, (Horse) Q9GKR0, (Dog) Q9MYV3, (Rat) P16612, (Mouse) Q6GT23
Entrez Gene ID: (Bovine) 281572, (Human) 7422, (Chicken) 395909, (Horse) 100033839, (Dog) 403802, (Rat) 83785, (Guinea pig) 100713483, (Mouse) 22339
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.