Novus Biologicals
Manufacturer Code:NBP298380C
Catalog # NB082315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant VDR/NR1I1/Vitamin D Receptor (P48281 Met1-Ser422). |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,25-dihydroxyvitamin D3 receptor; NR1I1Nuclear receptor subfamily 1 group I member 1125-dihydroxyvitamin D3 receptor vitamin D125- dihydroxyvitamin D3 receptor vitamin D3 receptor; Nuclear receptor subfamily 1 group I member 1; protein phosphatase 1, regulatory subunit 163; VDR; vitamin D nuclear receptor variant 1; vitamin D receptor; Vitamin D3 receptor
Gene Aliases: NR1I1; PPP1R163; VDR
UniProt ID: (Human) P11473
Entrez Gene ID: (Human) 7421
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.