Invitrogen
Manufacturer Code:PA5109236
Catalog # PIPA5109236
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,25-dihydroxyvitamin D3 receptor; 125-dihydroxyvitamin D3 receptor HGNC:12679 I79_025795 Nr1i1 Nuclear receptor subfamily 1 group I member 1 PPP1R163 Vdr Vitamin D3 receptor; Nuclear receptor subfamily 1 group I member 1; protein phosphatase 1, regulatory subunit 163; VDR; vitamin D nuclear receptor variant 1; vitamin D receptor; Vitamin D3 receptor
Gene Aliases: NR1I1; PPP1R163; VDR
UniProt ID: (Human) P11473
Entrez Gene ID: (Human) 7421
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.