Novus Biologicals
Manufacturer Code:NBP255590
Catalog # NBP255590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DNSLESYMKEKLKQLIEKRIGKENFIEKLGFISKHELYSRASQKPQPNFPSPEYMIFDHEFTKLVKELEGVITKAIYKSSEE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: General vesicular transport factor p115; p115 Transcytosis-associated protein USO1 homolog vesicle docking protein (yeast) USO1 vesicle docking protein homolog (yeast) vesicle docking protein vesicle docking protein p115; Protein USO1 homolog; TAP; transcytosis associated protein; Transcytosis-associated protein; USO1 vesicle docking protein homolog; vesicle docking protein p115; Vesicle-docking protein
Gene Aliases: P115; TAP; USO1; VDP
UniProt ID: (Human) O60763
Entrez Gene ID: (Human) 8615
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.