Novus Biologicals
Manufacturer Code:NBP217245
Catalog # NBP217245
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human VDP/p115. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: General vesicular transport factor p115; p115 Transcytosis-associated protein USO1 homolog vesicle docking protein (yeast) USO1 vesicle docking protein homolog (yeast) vesicle docking protein vesicle docking protein p115; Protein USO1 homolog; TAP; transcytosis associated protein; Transcytosis-associated protein; USO1 vesicle docking protein homolog; vesicle docking protein p115; Vesicle-docking protein
Gene Aliases: P115; TAP; USO1; VDP
UniProt ID: (Human) O60763
Entrez Gene ID: (Human) 8615
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.