Novus Biologicals
Manufacturer Code:NBP294176100UL
Catalog # NB022909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human VDAC3 (NP_005653.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HD-VDAC3 hVDAC3 Outer mitochondrial membrane protein porin 3 VDAC-3 voltage-dependent anion channel 3 voltage-dependent anion-selective channel protein 3; Outer mitochondrial membrane protein porin 3; VDAC-3; voltage-dependent anion channel 3; Voltage-dependent anion-selective channel protein 3
Gene Aliases: HD-VDAC3; VDAC-3; VDAC3
UniProt ID: (Human) Q9Y277
Entrez Gene ID: (Human) 7419
Molecular Function:
anion channel
ion channel
transporter
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.