Invitrogen
Manufacturer Code:PA555444
Catalog # PIPA555444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Human VDAC2/VDAC3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Store at &minus20°C long term. Avoid freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Outer mitochondrial membrane protein porin 2; Outer mitochondrial membrane protein porin 3; VDAC-2; VDAC-3; VDAC2 VDAC3 RP11-375G3.1 FLJ23841 Voltage-dependent anion-selective channel protein 2 Voltage-dependent anion-selective channel protein 3 hVDAC-2 hVDAC-3; voltage-dependent anion channel 2; voltage-dependent anion channel 3; Voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 3
Gene Aliases: HD-VDAC3; POR; VDAC-3; VDAC2; VDAC3
UniProt ID: (Human) P45880, (Human) Q9Y277
Entrez Gene ID: (Human) 7417, (Human) 7419
Molecular Function: anion channel ion channel transporter voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.