Invitrogen
Manufacturer Code:PA5116887
Catalog # PIPA5116887
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VDAC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B36-VDAC; B36-VDAC FLJ23841 mVDAC2 mVDAC6 Outer mitochondrial membrane protein porin 2 POR RP11-375G3.1 Vdac2 VDAC-2 VDAC2 protein Vdac6 VDAC-6 voltage dependent anion channel 2 voltage-dependent anion channel 2 voltage-dependent anion-selective channel protein 2 Voltage-dependent anion-selective channel protein 6; Outer mitochondrial membrane protein porin 2; VDAC-2; VDAC-6; voltage-dependent anion channel 2; Voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 6
Gene Aliases: mVDAC2; mVDAC6; POR; VDAC2; Vdac6
UniProt ID: (Human) P45880, (Mouse) Q60930, (Rat) P81155
Entrez Gene ID: (Human) 7417, (Mouse) 22334, (Rat) 83531
Molecular Function: anion channel ion channel transporter voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.