Abnova
Manufacturer Code:PAB0382
Catalog # 89-114-193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 120-132 of human VDAC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of VDAC2.
For Research Use Only
Protein Aliases: FLJ23841; Outer mitochondrial membrane protein porin 2; VDAC-2; voltage-dependent anion channel 2; Voltage-dependent anion-selective channel protein 2
Gene Aliases: POR; VDAC2
UniProt ID: (Human) P45880
Entrez Gene ID: (Human) 7417
Molecular Function: anion channel ion channel transporter voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.