Proteintech
Manufacturer Code:116631AP150UL
Catalog # 50-172-7355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VDAC2 Fusion Protein Ag2266 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B36-VDAC; hVDAC2 VDAC 2 VDAC2; Outer mitochondrial membrane protein porin 2; VDAC-2; VDAC-6; voltage-dependent anion channel 2; Voltage-dependent anion-selective channel protein 2; Voltage-dependent anion-selective channel protein 6
Gene Aliases: mVDAC2; mVDAC6; POR; VDAC2; Vdac6
UniProt ID: (Human) P45880, (Mouse) Q60930, (Rat) P81155
Entrez Gene ID: (Human) 7417, (Mouse) 22334, (Rat) 83531
Molecular Function:
anion channel
ion channel
transporter
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.