Invitrogen
Manufacturer Code:PA561942
Catalog # PIPA561942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human VCX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Variable charge protein on X with ten repeats; Variable charge X-linked protein 1; variable charge, X chromosome; Variably charged protein X-B1; VCX-10r; VCX-10r VCX-B1 VCX1 VCX10R VCXB1 variable charge X-linked protein 1 variable charge protein on X with ten repeats variable charge X chromosome variably charged protein X-B1; VCX-B1
Gene Aliases: VCX; VCX-10r; VCX-B1; VCX1; VCX10R; VCXB1
UniProt ID: (Human) Q9H320
Entrez Gene ID: (Human) 26609
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.