Invitrogen
Manufacturer Code:PA585230
Catalog # PIPA585230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human VCPIP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating protein VCPIP1; DKFZp686G038 antibody DUBA3 antibody FLJ23132 antibody FLJ60694 antibody KIAA1850 antibody VCIP135 antibody VCPIP1 antibody valosin-containing protein (p97)/p47 complex-interacting protein p135 antibody deubiquitinating protein VCIP135 antibody valosin-containing protein p97/p47 complex-interacting protein 1 antibody valosin-containing protein p97/p47 complex-interacting protein p135 antibody valosin containing protein (p97)/p47 complex interacting protein 1 antibody; valosin containing protein (p97)/p47 complex interacting protein 1; valosin-containing protein (p97)/p47 complex-interacting protein p135; Valosin-containing protein p97/p47 complex-interacting protein 1; Valosin-containing protein p97/p47 complex-interacting protein p135; VCP/p47 complex-interacting 135-kDa protein
Gene Aliases: 4932442A08; 5730421J18Rik; 5730538E15Rik; DUBA3; KIAA1850; mKIAA1850; VCIP135; VCPIP1
UniProt ID: (Human) Q9H5R8, (Mouse) Q8CDG3
Entrez Gene ID: (Human) 80124, (Mouse) 70675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.