Abnova
Manufacturer Code:H00080124A01
Catalog # 89-107-357
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1018 a.a. &bsim 1114 a.a) partial recombinant protein with GST tag.,VCPIP1 (NP_079330 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant VCPIP1.
For Research Use Only
Protein Aliases: Deubiquitinating protein VCPIP1; DKFZp686G038; DUBA3; FLJ23132; FLJ60694; KIAA1850; valosin containing protein (p97)/p47 complex interacting protein 1; valosin-containing protein (p97)/p47 complex-interacting protein p135; Valosin-containing protein p97/p47 complex-interacting protein 1; Valosin-containing protein p97/p47 complex-interacting protein p135; VCIP135; VCP/p47 complex-interacting 135-kDa protein
Gene Aliases: DUBA3; KIAA1850; VCIP135; VCPIP1
UniProt ID: (Human) Q9H5R8
Entrez Gene ID: (Human) 80124
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.