Proteintech
Manufacturer Code:178021AP150UL
Catalog # 50-173-1801
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VCPIP1 Fusion Protein Ag12035 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deubiquitinating protein VCIP135; Deubiquitinating protein VCPIP1; DUBA3 KIAA1850 VCIP135 VCPIP1; valosin containing protein (p97)/p47 complex interacting protein 1; valosin-containing protein (p97)/p47 complex-interacting protein 135; valosin-containing protein (p97)/p47 complex-interacting protein p135; Valosin-containing protein p97/p47 complex-interacting protein 1; Valosin-containing protein p97/p47 complex-interacting protein p135; VCP(p97)/p47-interacting protein; VCP/p47 complex-interacting 135-kDa protein
Gene Aliases: 4932442A08; 5730421J18Rik; 5730538E15Rik; DUBA3; KIAA1850; mKIAA1850; VCIP135; VCPIP1
UniProt ID: (Human) Q9H5R8, (Rat) Q8CF97, (Mouse) Q8CDG3
Entrez Gene ID: (Human) 80124, (Rat) 286761, (Mouse) 70675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.