Invitrogen
Manufacturer Code:PA586653
Catalog # PIPA586653
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 320-370 of Human VCP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15S Mg(2+) ATPase p97 subunit ATPase p97 IBMPFD MGC131997 MGC148092 MGC8560 TER ATPase TERA p97 transitional endoplasmic reticulum ATPase valosin-containing protein yeast Cdc48p homolog; 15S Mg(2+)-ATPase p97 subunit; AAA ATPase p97; epididymis luminal protein 220; epididymis secretory protein Li 70; homolog of yeast cdc48; Inv protein; TER ATPase; Transitional endoplasmic reticulum ATPase; Valosin-containing protein; VCP; yeast Cdc48p homolog
Gene Aliases: 3110001E05; ALS14; CDC48; CMT2Y; HEL-220; HEL-S-70; IBMPFD; IBMPFD1; p97; p97/VCP; TERA; VCP
UniProt ID: (Human) P55072, (Mouse) Q6PI18, (Rat) P46462
Entrez Gene ID: (Human) 7415, (Mouse) 269523, (Rat) 116643
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.