Novus Biologicals
Manufacturer Code:NBP132663
Catalog # NBP132663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human VCPIP1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Deubiquitinating protein VCPIP1; DKFZp686G038 DUBA3 EC 3.4.22.- FLJ23132 KIAA1850FLJ60694 valosin containing protein (p97)/p47 complex interacting protein 1 valosin-containing protein (p97)/p47 complex-interacting protein p135 Valosin-containing protein p97/p47 complex-interacting protein 1 Valosin-containing protein p97/p47 complex-interacting protein p135 VCIP135deubiquitinating protein VCIP135 VCPIP1 VCIP135; valosin containing protein (p97)/p47 complex interacting protein 1; valosin-containing protein (p97)/p47 complex-interacting protein p135; Valosin-containing protein p97/p47 complex-interacting protein 1; Valosin-containing protein p97/p47 complex-interacting protein p135; VCP/p47 complex-interacting 135-kDa protein
Gene Aliases: DUBA3; KIAA1850; VCIP135; VCPIP1
UniProt ID: (Human) Q9H5R8
Entrez Gene ID: (Human) 80124
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.