Abnova
Manufacturer Code:MAB9799
Catalog # 89-113-791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | STA |
Immunogen | Native purified VCAM1 from human DS6 T cell line. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. |
Mouse monoclonal antibody raised against native VCAM1.
For Research Use Only
Protein Aliases: CD106; CD106 antigen; DKFZp779G2333; INCAM-100; MGC99561; V-CAM 1; Vascular cell adhesion protein 1
Gene Aliases: CD106; INCAM-100; VCAM1
UniProt ID: (Human) P19320
Entrez Gene ID: (Human) 7412
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.