R&D Systems
Manufacturer Code:MAB6434
Catalog # MAB6434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 112702 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse VCAM-1 Phe25-Glu698 Accession # P29533 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD106; CD106 antigen; CD106 CD106 antigen DKFZp779G2333 INCAM-100 L1CAM MGC99561 vascular cell adhesion molecule 1 vascular cell adhesion protein 1 V-CAM 1 VCAM1 VCAM-1; INCAM-100; V-CAM 1; Vascular cell adhesion protein 1
Gene Aliases: CD106; INCAM-100; VCAM1
UniProt ID: (Human) P19320
Entrez Gene ID: (Human) 7412
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.