Invitrogen
Manufacturer Code:PA580214
Catalog # PIPA580214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat VCAM1 recombit protein (Position: F25-L270). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD106; V-CAM 1; Vascular cell adhesion protein 1; Vascular cell adhesion protein 1 V-CAM 1 VCAM-1 CD106 Vcam1 Vcam-1; VCAM-1
Gene Aliases: CD106; Vcam-1; Vcam1; VCAM1B
UniProt ID: (Rat) P29534
Entrez Gene ID: (Rat) 25361, (Mouse) 22329
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.