Invitrogen
Manufacturer Code:MA526008
Catalog # PIMA526008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A3 |
Immunogen | Full length human recombit protein of VBP1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HIBBJ46; Prefoldin subunit 3; RP13-228J13.4 PFD3 PFDN3 VBP-1 HIBBJ46 VHL binding protein-1 VHL-binding protein 1 prefoldin 3 prefoldin subunit 3 von Hippel-Lindau-binding protein 1; VBP-1; VHL binding protein-1; VHL-binding protein 1; von Hippel-Lindau binding protein 1; von Hippel-Lindau-binding protein 1
Gene Aliases: EGK_21124; HIBBJ46; PFD3; PFDN3; VBP-1; VBP1
UniProt ID: (Human) P61758, (Mouse) P61759
Entrez Gene ID: (Rhesus monkey) 703257, (Human) 7411, (Dog) 481089, (Mouse) 22327, (Rat) 681825
Molecular Function:
membrane traffic protein
vesicle coat protein
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.