Invitrogen
Manufacturer Code:PA599508
Catalog # PIPA599508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human VAV1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: oncogene vav p95 p95vav Proto-oncogene vav VAV vav 1 guanine nucleotide exchange factor vav 1 oncogene vav guanine nucleotide exchange factor 1 vav oncogene vav proto-oncogene VAV1 vav-T; p95; p95vav; Proto-oncogene vav; vav 1 guanine nucleotide exchange factor; vav 1 oncogene; vav oncogene
Gene Aliases: VAV; vav-T; VAV1
UniProt ID: (Human) P15498, (Mouse) P27870
Entrez Gene ID: (Human) 7409, (Rat) 25156, (Mouse) 22324
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.