Novus Biologicals
Manufacturer Code:NBP274831PEATT594
Catalog # N274831P594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H3 |
Immunogen | Full length recombinant protein of human VAT1L (NP_065978) produced in HEK293T cells. |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: KIAA1576synaptic vesicle membrane protein VAT-1 homolog-like vesicle amine transport protein 1 homolog (T. californica)-like vesicle amine transport protein 1 homolog-like (T. californica); Synaptic vesicle membrane protein VAT-1 homolog-like; vesicle amine transport protein 1 homolog (T. californica)-like; vesicle amine transport protein 1 homolog-like (T. californica)
Gene Aliases: KIAA1576; VAT1L
UniProt ID: (Human) Q9HCJ6
Entrez Gene ID: (Human) 57687
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.