Invitrogen
Manufacturer Code:PA571863
Catalog # PIPA571863
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 216-245 amino acids from the C-terminal region of human VASH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: testicular tissue protein Li 222; Tubulinyl-Tyr carboxypeptidase 2; Vasohibin-2; Vasohibin-2 Vasohibin-like protein VASH2 VASHL; Vasohibin-like protein
Gene Aliases: B130052G07Rik; VASH2; VASHL
UniProt ID: (Human) Q86V25, (Mouse) Q8BRA1
Entrez Gene ID: (Human) 79805, (Mouse) 226841
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.