Novus Biologicals
Manufacturer Code:NBP29305920UL
Catalog # NB028012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 864-1063 of human VARS2 (NP_065175.4). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZP434L1435 EC 6.1.1 EC 6.1.1.9 Em:AB023048.4 KIAA1885MGC138259 MGC142165 valine tRNA ligase 2 mitochondrial (putative) Valine--tRNA ligase valRS valyl-tRNA synthetase 2 mitochondrial (putative) valyl-tRNA synthetase 2-like valyl-tRNA synthetase like valyl-tRNA synthetase mitochondrial Valyl-tRNA synthetase-like VARS2L VARSL; valine tRNA ligase 2, mitochondrial (putative); Valine--tRNA ligase, mitochondrial; ValRS; Valyl-tRNA synthetase; valyl-tRNA synthetase like; Valyl-tRNA synthetase-like
Gene Aliases: COXPD20; KIAA1885; VALRS; VARS2; VARS2L; VARSL
UniProt ID: (Human) Q5ST30
Entrez Gene ID: (Human) 57176
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.