Proteintech
Manufacturer Code:157761AP150UL
Catalog # 50-173-0470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VARS2 Fusion Protein Ag8460 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1885 Valine tRNA ligase ValRS Valyl tRNA synthetase like VARS2 VARS2L VARSL; valine tRNA ligase 2, mitochondrial (putative); Valine--tRNA ligase, mitochondrial; ValRS; Valyl-tRNA synthetase; valyl-tRNA synthetase 2, mitochondrial (putative); valyl-tRNA synthetase like; valyl-tRNA synthetase, mitochondrial; Valyl-tRNA synthetase-like
Gene Aliases: 1190004I24Rik; COXPD20; KIAA1885; mKIAA1885; VALRS; VARS2; VARS2L; VARSL
UniProt ID: (Human) Q5ST30, (Mouse) Q3U2A8, (Rat) Q6MG21
Entrez Gene ID: (Human) 57176, (Mouse) 68915, (Rat) 309596
Molecular Function: aminoacyl-tRNA synthetase ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.