Invitrogen
Manufacturer Code:PA576475
Catalog # PIPA576475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human VARS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G7A VARS1 VARS2; Protein G7a; valine tRNA ligase 1, cytoplasmic; Valine--tRNA ligase; ValRS; Valyl-tRNA synthetase; valyl-tRNA synthetase 2
Gene Aliases: Bat6; D17H6S56E; G7A; TrsVal; VARS; VARS1; VARS2
UniProt ID: (Human) P26640, (Rat) Q04462, (Mouse) Q9Z1Q9
Entrez Gene ID: (Human) 7407, (Rat) 25009, (Mouse) 22321
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.