Invitrogen
Manufacturer Code:PA5113815
Catalog # PIPA5113815
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human VARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bat6 D17H6S56E EC 6.1.1.9 G7A Protein G7a TrsVal valine tRNA ligase 1 cytoplasmic Valine--tRNA ligase ValRS valyl trna synthetase Valyl-tRNA synthetase valyl-tRNA synthetase 2 VARS VARS1 VARS2; Protein G7a; valine tRNA ligase 1, cytoplasmic; Valine--tRNA ligase; ValRS; Valyl-tRNA synthetase; valyl-tRNA synthetase 2
Gene Aliases: G7A; VARS; VARS1; VARS2
UniProt ID: (Human) P26640
Entrez Gene ID: (Human) 7407
Molecular Function:
aminoacyl-tRNA synthetase
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.