Invitrogen
Manufacturer Code:PA522188
Catalog # PIPA522188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 29 and 122 of VAPA (Uniprot IDNo.Q9P0L0) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 33 kDa VAMP-associated protein; synaptobrevin-associated protein of 33kDa; VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa; VAMP-A; VAMP-associated protein A; VAP-33; VAP-33 VAP-A VAP33 hVAP-33 33 kDa VAMP-associated protein VAMP-A VAMP-associated protein A vesicle-associated membrane protein-associated protein A; VAP-A; Vesicle-associated membrane protein-associated protein A
Gene Aliases: 33kDa; hVAP-33; VAP-33; VAP-A; VAP33; VAPA
UniProt ID: (Human) Q9P0L0, (Mouse) Q9WV55
Entrez Gene ID: (Human) 9218, (Mouse) 30960
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.