Novus Biologicals
Manufacturer Code:NBP289625AF594
Catalog # NB108532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 103 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human VAP-B (O95292-1 Met1-Pro132). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ALS8VAMP-B/VAMP-C VAMP (vesicle-associated membrane protein)-associated protein B and C VAMP-associated 33 kDa protein VAMP-associated protein B/C VAMP-B VAP-B VAP-B/VAP-C VAPC VAP-C vesicle-associated membrane protein-associated protein B/C; VAMP (vesicle-associated membrane protein)-associated protein B and C; VAMP-associated 33 kDa protein; VAMP-B/VAMP-C; Vesicle-associated membrane protein-associated protein B/C
Gene Aliases: ALS8; UNQ484/PRO983; VAMP-B; VAP-B; VAPB
UniProt ID: (Human) O95292
Entrez Gene ID: (Human) 9217
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.