Novus Biologicals
Manufacturer Code:NBP274825AF405
Catalog # N274825A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10E10 |
Immunogen | Recombinant protein fragment corresponding to amino acids 2-227 of human VAP-A (NP_919415) produced in E.coli. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 33 kDa VAMP-associated protein; hVAP-33 MGC374533 kDa VAMP-associated protein VAMP (vesicle-associated membrane protein)-associated protein A 33kDa VAMP-A VAP-33 VAP33VAMP (vesicle-associated membrane protein)-associated protein A (33kD) VAP-AVAMP-associated protein A vesicle-associated membrane protein-associated protein A; VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa; VAMP-A; VAMP-associated protein A; VAP-33; Vesicle-associated membrane protein-associated protein A
Gene Aliases: hVAP-33; VAP-33; VAP-A; VAP33; VAPA
UniProt ID: (Human) Q9P0L0
Entrez Gene ID: (Human) 9218
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.