Novus Biologicals
Manufacturer Code:NBP131237
Catalog # NBP131237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human VAPA. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 33 kDa VAMP-associated protein; hVAP-33 MGC374533 kDa VAMP-associated protein VAMP (vesicle-associated membrane protein)-associated protein A 33kDa VAMP-A VAP-33 VAP33VAMP (vesicle-associated membrane protein)-associated protein A (33kD) VAP-AVAMP-associated protein A vesicle-associated membrane protein-associated protein A; VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa; VAMP-A; VAMP-associated protein A; VAP-33; Vesicle-associated membrane protein-associated protein A
Gene Aliases: hVAP-33; VAP-33; VAP-A; VAP33; VAPA
UniProt ID: (Human) Q9P0L0
Entrez Gene ID: (Human) 9218
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.