Novus Biologicals
Manufacturer Code:NBP281043AF700
Catalog # NB014703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | TK8-14 |
Immunogen | Affinity purified VAP-1/AOC3 from tonsil stroma of human origin. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: amine oxidase copper containing 3 (vascular adhesion protein 1) Copper amine oxidase EC 1.4.3 HPAOSSAO Semicarbazide-sensitive amine oxidase VAP1EC 1.4.3.21 VAP-1membrane primary amine oxidase Vascular adhesion protein 1; amine oxidase, copper containing 3 (vascular adhesion protein 1); Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; Semicarbazide-sensitive amine oxidase; SSAO; VAP-1; Vascular adhesion protein 1
Gene Aliases: AOC3; HPAO; SSAO; VAP-1; VAP1
UniProt ID: (Human) Q16853
Entrez Gene ID: (Human) 8639
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.