R&D Systems
Manufacturer Code:IC39571S100UG
Catalog # IC39571S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 393106 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human VAP-1/AOC3 Gly27-Asn763 Accession # Q16853 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: amine oxidase copper containing 3 (vascular adhesion protein 1) AOC3 Copper amine oxidase EC 1.4.3 HPAO HPAOSSAO Semicarbazide-sensitive amine oxidase SSAO VAP1 VAP1EC 1.4.3.21 VAP-1membrane primary amine oxidase Vascular adhesion protein 1; amine oxidase, copper containing 3 (vascular adhesion protein 1); Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; Semicarbazide-sensitive amine oxidase; SSAO; VAP-1; Vascular adhesion protein 1
Gene Aliases: AOC3; HPAO; SSAO; VAP-1; VAP1
UniProt ID: (Human) Q16853
Entrez Gene ID: (Human) 8639
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.