Novus Biologicals
Manufacturer Code:NBP131935
Catalog # NBP131935
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human VAP1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: amine oxidase copper containing 3 (vascular adhesion protein 1) Copper amine oxidase EC 1.4.3 HPAOSSAO Semicarbazide-sensitive amine oxidase VAP1EC 1.4.3.21 VAP-1membrane primary amine oxidase Vascular adhesion protein 1; amine oxidase, copper containing 3 (vascular adhesion protein 1); Copper amine oxidase; HPAO; Membrane primary amine oxidase; placenta copper monamine oxidase; Semicarbazide-sensitive amine oxidase; SSAO; VAP-1; Vascular adhesion protein 1
Gene Aliases: AOC3; HPAO; SSAO; VAP-1; VAP1
UniProt ID: (Human) Q16853
Entrez Gene ID: (Human) 8639
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.