Invitrogen
Manufacturer Code:PA5113503
Catalog # PIPA5113503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Vang-like protein 2 protein (1-110AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C530001F03Rik KIAA1215 loop tail associated protein loop-tail Loop-tail protein 1 Loop-tail protein 1 homolog Loop-tail-associated protein Lp Lpp1 Ltap MGC119403 MGC119404 RGD:1309442} ska<m17Jus> ska17 STB1 stbm STBM1 strabismus Strabismus 1 Van Gogh-like protein 2 vang vang (van gogh)-like 2 Vang1l2 VANGL planar cell polarity protein 2 Vangl2 vangl2 {ECO:0000312 vang-like 2 (van gogh Drosophila) Vang-like protein 2; loop tail associated protein; Loop-tail protein 1; Loop-tail protein 1 homolog; Loop-tail-associated protein; Strabismus 1; Van Gogh-like protein 2; vang (van gogh)-like 2; vang-like 2 (van gogh, Drosophila); Vang-like protein 2
Gene Aliases: C530001F03Rik; KIAA1215; loop-tail; Lp; LPP1; LTAP; ska17; ska<m17Jus>; STB1; STBM; STBM1; strabismus; Vang1l2; VANGL2
UniProt ID: (Human) Q9ULK5, (Mouse) Q91ZD4
Entrez Gene ID: (Human) 57216, (Mouse) 93840
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.