Abnova
Manufacturer Code:H00057216K
Catalog # 89-200-518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human VANGL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human VANGL2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: KIAA1215 LPP1 LTAP MGC119403 MGC119404 STB1 STBM STBM1; Loop-tail protein 1 homolog; loop-tail-associated protein; Strabismus 1; Van Gogh-like protein 2; vang-like 2 (van gogh, Drosophila); Vang-like protein 2
Gene Aliases: KIAA1215; LPP1; LTAP; STB1; STBM; STBM1; VANGL2
UniProt ID: (Human) Q9ULK5
Entrez Gene ID: (Human) 57216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.