Invitrogen
Manufacturer Code:MA537661
Catalog # PIMA537661
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1414CT354.12.23.93 |
Immunogen | KLH conjugated synthetic peptide between 2~24 amino acids from the N-terminal region of human VAMP8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AU041171 EDB Endobrevin vamp VAMP8 VAMP-8 vesicle associated membrane protein 8 vesicle-associated membrane protein 8 vesicle-associated membrane protein 8 (endobrevin); EDB; Endobrevin; VAMP-8; Vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin)
Gene Aliases: EDB; VAMP-8; VAMP8
UniProt ID: (Human) Q9BV40
Entrez Gene ID: (Human) 8673
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.