Invitrogen
Manufacturer Code:PA570439
Catalog # PIPA570439
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of rat VAMP8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EDB; Endobrevin; VAMP-8; Vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin)
Gene Aliases: AU041171; EDB; endobrevin; VAMP-8; VAMP8; wu:fa18d07; zgc:56588
UniProt ID: (Bovine) Q3T0Y8, (Human) Q9BV40, (Mouse) O70404, (Rat) Q9WUF4
Entrez Gene ID: (Horse) 100053197, (Bovine) 507309, (Dog) 609784, (Rabbit) 100338966, (Human) 8673, (Mouse) 22320, (Rat) 83730, (Guinea pig) 100728978, (Zebrafish) 327007
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.