Proteintech
Manufacturer Code:155461AP150UL
Catalog # 50-173-0304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VAMP8 Fusion Protein Ag7903 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EDB; EDB Endobrevin VAMP 8 VAMP8; Endobrevin; VAMP-8; Vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin)
Gene Aliases: AU041171; EDB; endobrevin; VAMP-8; VAMP8
UniProt ID: (Human) Q9BV40, (Mouse) O70404, (Rat) Q9WUF4
Entrez Gene ID: (Human) 8673, (Mouse) 22320, (Rat) 83730
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.