Invitrogen
Manufacturer Code:PA595373
Catalog # PIPA595373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human SYBL1 recombit protein (Position: N24-T190). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: synaptobrevin-like 1; Synaptobrevin-like protein 1; tetanus neurotoxin-insensitive VAMP; Tetanus-insensitive VAMP; Ti-VAMP; VAMP-7; Vesicle-associated membrane protein 7; Vesicle-associated membrane protein 7 VAMP-7 Synaptobrevin-like protein 1 Tetanus-insensitive VAMP Ti-VAMP VAMP7 SYBL1
Gene Aliases: SYBL1; TI-VAMP; TIVAMP; VAMP-7; VAMP7
UniProt ID: (Human) P51809, (Rat) Q9JHW5
Entrez Gene ID: (Human) 6845, (Rat) 85491
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.