Invitrogen
Manufacturer Code:OSS00047G
Catalog # PIOSS00047G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from aa region 50-100 of human VAMP7 conjugated to an immunogenic carrier protein was used as the antigen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: synaptobrevin-like 1; Synaptobrevin-like protein 1; tetanus neurotoxin-insensitive VAMP; Tetanus-insensitive VAMP; Ti-VAMP; VAMP-7; Vesicle-associated membrane protein 7; Vesicle-associated membrane protein 7 VAMP7 SYBL1 VAMP-7 Synaptobrevin-like protein 1 Tetanus-insensitive VAMP Ti-VAMP
Gene Aliases: SYBL1; TI-VAMP; TIVAMP; VAMP-7; VAMP7
UniProt ID: (Human) P51809, (Mouse) P70280, (Rat) Q9JHW5
Entrez Gene ID: (Human) 6845, (Mouse) 20955, (Rat) 85491
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.