Invitrogen
Manufacturer Code:MA524435
Catalog # PIMA524435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D4 |
Immunogen | VAMP5 (NP&lowbar006625 1 a.a. &bsim 70 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Myobrevin; Myobrevin VAMP-5 VAMP5 Vesicle-associated membrane protein 5; VAMP-5; Vesicle-associated membrane protein 5
Gene Aliases: HSPC191; VAMP5
UniProt ID: (Human) O95183
Entrez Gene ID: (Human) 10791
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.