Abnova
Manufacturer Code:H00010791A01
Catalog # 89-106-409
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. &bsim 70 a.a) partial recombinant protein with GST tag.,VAMP5 (NP_006625 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant VAMP5.
For Research Use Only
Protein Aliases: Myobrevin; VAMP-5; Vesicle-associated membrane protein 5
Gene Aliases: HSPC191; VAMP5
UniProt ID: (Human) O95183
Entrez Gene ID: (Human) 10791
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.