Proteintech
Manufacturer Code:118221AP150UL
Catalog # 50-172-7482
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VAMP5 Fusion Protein Ag2390 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Myobrevin; Myobrevin VAMP 5 VAMP5; VAMP-5; Vesicle-associated membrane protein 5
Gene Aliases: AF119384; Camp; HSPC191; VAMP5
UniProt ID: (Human) O95183, (Mouse) Q9Z2P8, (Rat) Q9Z2J5
Entrez Gene ID: (Human) 10791, (Mouse) 53620, (Rat) 89818
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.