Proteintech
Manufacturer Code:107381AP150UL
Catalog # 50-172-6554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VAMP4 Fusion Protein Ag1090 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: VAMP 4 VAMP24 VAMP4; VAMP-4; vesicle associated membrane protein 4; Vesicle-associated membrane protein 4
Gene Aliases: D1Ertd147e; VAMP-4; VAMP24; VAMP4
UniProt ID: (Human) O75379, (Mouse) O70480
Entrez Gene ID: (Human) 8674, (Mouse) 53330, (Rat) 364033
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.