Abnova
Manufacturer Code:MAB2061
Catalog # 89-110-802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AT4G9 |
Immunogen | Recombinant protein corresponding to amino acids 1-77 of human VAMP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant VAMP3.
For Research Use Only
Protein Aliases: CEB; Cellubrevin; Synaptobrevin-3; VAMP-3; Vesicle-associated membrane protein 3
Gene Aliases: CEB; SYB3; VAMP3
UniProt ID: (Human) Q15836
Entrez Gene ID: (Human) 9341
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.