Invitrogen
Manufacturer Code:PA5116218
Catalog # PIPA5116218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 15-43 amino acids from the Central region of human VAMP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CEB; CEB Cellubrevin D130027G05Rik Syb3 Synaptobrevin-3 vamp Vamp3 VAMP-3 vesicle associated membrane protein 3 vesicle-associated membrane protein 3; Cellubrevin; Synaptobrevin-3; VAMP-3; Vesicle-associated membrane protein 3
Gene Aliases: CEB; D130027G05Rik; SYB3; VAMP-3; VAMP3; zgc:86726
UniProt ID: (Human) Q15836, (Mouse) P63024
Entrez Gene ID: (Human) 9341, (Mouse) 22319, (Zebrafish) 415163
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.