Invitrogen
Manufacturer Code:PA585779
Catalog # PIPA585779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse VAMP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Syb2 antibody VAMP-2 antibody VAMP2 antibody VAMP2 antibody sybII antibody vesicle-associated membrane protein 2 antibody; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin II; Synaptobrevin-2; VAMP-2; vesicle-associated membrane 2; Vesicle-associated membrane protein (synaptobrevin 2); Vesicle-associated membrane protein 2
Gene Aliases: RATVAMPB; RATVAMPIR; SYB; Syb-2; Syb2; sybII; Vamp2
UniProt ID: (Mouse) P63044, (Rat) P63045
Entrez Gene ID: (Mouse) 22318, (Rat) 24803
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.