Invitrogen
Manufacturer Code:OSS00035W
Catalog # PIOSS00035W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from aa region 10-60 of human VAMP2 conjugated to an immunogenic carrier protein was used as the antigen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin II; Synaptobrevin-2; VAMP-2; VAMP-2 Synaptobrevin-2 VAMP2 SYB2; vesicle-associated membrane 2; Vesicle-associated membrane protein (synaptobrevin 2); Vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2)
Gene Aliases: RATVAMPB; RATVAMPIR; SYB; Syb-2; SYB2; sybII; VAMP-2; VAMP2
UniProt ID: (Human) P63027, (Mouse) P63044, (Rat) P63045
Entrez Gene ID: (Human) 6844, (Mouse) 22318, (Rat) 24803
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.