Novus Biologicals
Manufacturer Code:NBP232095
Catalog # NBP232095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 1 to 50 of human Vesicle-Associated Membrane Protein 8 using the numbering given in entry NP_003752.2 (GeneID 8673). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EDB; EDBVAMP-8 Endobrevin vesicle-associated membrane protein 8 vesicle-associated membrane protein 8 (endobrevin); Endobrevin; VAMP-8; Vesicle-associated membrane protein 8; vesicle-associated membrane protein 8 (endobrevin)
Gene Aliases: EDB; VAMP-8; VAMP8
UniProt ID: (Human) Q9BV40
Entrez Gene ID: (Human) 8673
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.